Learn More
CD25 Rat anti-Mouse, Biotin, Clone: 7D4, BD
Rat Monoclonal Antibody
$126.00 - $310.00
Specifications
Antigen | CD25 |
---|---|
Clone | 7D4 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
The 7D4 antibody reacts with CD25, the low affinity IL-2 Receptor (IL-2Rα, p55) expressed on activated T and B lymphocytes from all mouse strains tested. IL-2Rα by itself is not a signaling receptor. However, it can combine with IL-2 Receptor β (CD122) and γ (CD132) chains to form high-affinity, signaling receptor complexes for IL-2. Resting T and B lymphocytes and resting and activated NK cells do not express IL-2Rα. CD25 is transiently expressed at a low level during normal B-cell development in the bone marrow on the CD45R/B220low
TdT- sIg- Pre-B/Pre-B-II and CD45R/B220low TdT- sIgM+ sIgD- immature B stages, but not on the CD45R/B220low TdT+ sIg- Pro-B/Pre-B-I stage nor on CD45R/B220high TdTsIgM+ sIgD+ mature B cells. It is expressed at a higher level during a very early stage of T-cell development in fetal and adult thymus. Peripheral CD25+ CD4+ T lymphocytes called regulatory T (Treg) cells are involved in the maintenance of self-tolerance. It has also been reported that dendritic cells express CD25, recognized by mAb 7D4 (Cat. no. 553068). The
3C7 antibody recognizes an epitope of CD25 which is distinct from those recognized by mAbs 7D4 and PC61 (Cat. no. 553866), and it blocks binding of IL-2 to CD25. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 7D4
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: IL-2-dependent BALB/c mouse helper T-cell clone HT-2
Flow Cytometry
Specifications
CD25 | |
0.5mg/mL | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgM κ | |
Affinity Purified | |
IL-2 Receptor α chain, p55 |
7D4 | |
Flow Cytometry | |
Biotin | |
RUO | |
Mouse | |
IL-2-dependent BALB/c mouse helper T-cell clone HT-2 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.