Learn More
CD25 Mouse anti-Rat, Biotin, Clone: OX-39, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559981
Description
The OX-39 antibody reacts with the α chain of the IL-2 receptor on T lymphoblasts and thymic and splenic dendritic cells. CD25 has also been detected on rat intestinal epithelial cells. It has been reported that OX-39 mAb weakly blocks binding of IL-2 to T-cell blasts and that it blocks IL-2 stimulated epithelial cell migration in an in vitro model of wound healing.

Specifications
CD25 | |
Monoclonal | |
0.5mg/mL | |
IL-2R α Chain | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG1 κ |
Flow Cytometry | |
OX-39 | |
Biotin | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat T blasts from mixed-lymphocyte reactions | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.