Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25 (IL-2 Receptor α chain) Mouse anti-Human, Unlabeled, Clone: 2A3, BD

Catalog No. BDB347640 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347640 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347640 Supplier BD Biosciences Supplier No. 347640
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD25, clone 2A3, is derived from hybridization of mouse myeloma cell line NS-1 myeloma cells with spleen cells from BALB/c mice immunized with PHA-activated human T lymphocytes.

CD25 (Anti–IL-2R) recognizes an antigen that is the human low-affinity interleukin-2 receptor (IL-2R). The antigen has a molecular weight of 55 kilodaltons (kd).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25 (IL-2 Receptor α chain)
Applications Flow Cytometry
Classification Monoclonal
Clone 2A3
Concentration 5μg/mL
Conjugate Unconjugated
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Phytohemagglutinin-activated T Cells
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.