Learn More
CD252 Mouse anti-Human, PE, Clone: IK-1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 558164
Description
The ik-1 monoclonal antibody specifically binds to OX40 Ligand (OX40L) which is also known as CD252 and CD134 Ligand. OX40L is encoded by TNFSF4 (Tumor necrosis factor (ligand) superfamily, member 4). OX40L a 34 kDa type II transmembrane glycoprotein that is expressed by dendritic cells, B cells, human umbilical vein endothelial cells (HUVEC) and other types of vascular endothelial cells. OX40L serves as a costimulatory ligand by binding to and signaling through OX40 (CD134) which is expressed on activated T cells. It also mediates adhesion of activated T cells or OX40-positive transformed cells to vascular endothelial cells. Monocyte-derived dendritic cells (MoDC) express OX40 Ligand after stimulation through CD40. Ligation of OX40 Ligand expressed by activated MoDC can significantly increase their cytokine production.
Specifications
| CD252 | |
| Monoclonal | |
| 0.2mg/mL | |
| CD252; OX40 Ligand; CD134 Ligand; CD134L; TNFSF4; GP34; TXGP1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| IK-1 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human OX40 Ligand Transfected Cell Line | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.