Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD253 Rat anti-Mouse, Unlabeled, Clone: N2B2, BD

Catalog No. BDB550320 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550320 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550320 Supplier BD Biosciences Supplier No. 550320
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The N2B2 antibody reacts with mouse TRAIL (TNF-Related Apoptosis-Inducing Ligand, also known as APO-2 ligand or Ly-81), a 40-kDa type-II transmembrane protein of the TNF family. This antibody recognizes mouse Trail -transfected 2PK-3 cells, but not the parental cell line. TRAIL selectively induces apoptosis in various transformed cells, but not in normal cells. Five receptors have been identified for human TRAIL, whereas only one receptor has been described for the mouse counterpart thus far. The pattern of expression of mouse TRAIL is controversial. TRAIL has been detected, with a rabbit polyclonal antibody, on the surface of activated CD4+ and CD8+ T lymphocytes, as well as on CD45R/B220+ lymphocytes, and transformed T- and B-cell lines. Similar results were obtained by western blot and mRNA analyses. However, these results could not be reproduced with the N2B2 mAb. TRAIL expression, detected with the N2B2 mAb, is induced on CD3- NK1.1+ cells after stimulation with IL-2 or IL-15. However, neither freshly isolated nor activated T cells showed TRAIL expression on the cell surface. Furthermore, TRAIL mediates NK-cell cytotoxicity, which is blocked by the N2B2 mAb. TRAIL has been renamed as CD253 recently.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD253
Applications Flow Cytometry
Classification Monoclonal
Clone N2B2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description TRAIL, APO-2L, Ly-81, TL2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Trail-transfected 2PK-3 cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.