Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD258 (LIGHT) Mouse anti-Human, Alexa Fluor 647, Clone: 115520, BD

Catalog No. BDB564374 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564374 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564374 Supplier BD Biosciences Supplier No. 564374
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 115520 monoclonal antibody specifically binds to CD258 which is also known as LIGHT (is homologous to lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes). CD258 is also referred to as HVEM-L (Herpes virus entry mediator ligand), and LT- γ. This type II transmembrane glycoprotein belongs to TNF superfamily; it is encoded by TNFSF14 (tumor necrosis factor ligand superfamily member 14). CD258 is expressed by activated T cells and cultured dendritic cells. CD258 can costimulate T cells and antigen presenting cells as well as trigger apoptosis by some cell types. CD258 binds to CD270/LIGHT-R/HVEM and to the Lymphotoxin β Receptor (LT β R). LIGHT reportedly also binds to the decoy receptor (DcR3/TR6).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD258 (LIGHT)
Applications Flow Cytometry
Classification Monoclonal
Clone 115520
Conjugate Alexa Fluor 647
Description TNFSF14; LIGHT; HVEML; LTg
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human LIGHT Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.