Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD26 Mouse anti-Rat, Unlabeled, Clone: OX-61, BD

Catalog No. BDB559639 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559639 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559639 Supplier BD Biosciences Supplier No. 559639

Mouse Monoclonal Antibody

The OX-61 antibody reacts with a type II transmembrane glycoprotein, CD26, which is a serine exoprotease identified as dipeptidyl/peptidase IV. Among various biological activities, rat CD26 binds fibronectin and collagen. Rat CD26 is involved in the costimulation of thymocyte proliferation in vitro, particularly the CD4-/CD8- subset, and is developmentally regulated on hematopoietic cells. Although mouse and human CD26 anchor ADA (adenosine deaminase) to cell membranes, rat CD26 does not function as an ADA-binding protein. Rat CD26 is expressed in lung endothelial cells, as well as in various epithelial cells. T cells express lower levels of CD26 than CD4-/CD8- thymocytes. The distribution of CD26 antigen in rat bone marrow cells is similar to that of human CD26. OX-61 monoclonal antibody stains CD4+, CD8+, and Ig+ lymphocytes, and the staining increases upon activation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD26
Applications Flow Cytometry
Classification Monoclonal
Clone OX-61
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat dendritic cells enriched from thoracic duct lymph
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.