Learn More
CD26 Rat anti-Mouse, FITC, Clone: H194-112, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559652
Description
The H194-112 monoclonal antibody specifically binds to CD26, which is also known as, Thymocyte-activating molecule (THAM), or dipeptidyl peptidase IV (DPP IV, Dpp4). CD26 is a ∽220- kDa dimer formed of identical type-II transmembrane core polypeptides which undergo variable post-translational modifications. It is a multi-functional molecule with both ectopeptidase and signal-transducing activities. Studies with specific DPP IV inhibitors suggest that the enzymatic activity is involved in the mediation of T-cell activation events. The expression of CD26 is developmentally regulated in the thymus. Resting lymphoid cells of the bone marrow and peripheral B and T lymphocytes express low levels of CD26; bone-marrow and peritoneal myeloid cells do not. CD26 is also found on epithelial cells in the kidney, liver, small intestine, and lung. Cross-linked H194-112 mAb induces proliferation of immature and mature thymocytes in the presence of either IL-1 plus IL-2 or PMA; addition of IL-2 or IL-4 to PMA further enhances the activation.

Specifications
CD26 | |
Monoclonal | |
0.5mg/mL | |
Cd26; Dpp4; dipeptidylpeptidase 4; THAM, DPP IV | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
H194-112 | |
FITC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Cell-surface molecules solubilized from BALB/c mouse thymocytes | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.