Learn More
CD27 Mouse anti-Human, APC-H7, Clone: M-T271, BD
Mouse Monoclonal Antibody
$174.00 - $373.50
Specifications
Antigen | CD27 |
---|---|
Clone | M-T271 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-H7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560223
|
BD Biosciences
560223 |
25 Tests |
Each for $174.00
|
|
|||||
BDB560222
|
BD Biosciences
560222 |
100 Tests |
Each for $373.50
|
|
|||||
Description
Reacts with a disulphide-linked 110kDa dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. CD27 is highly induced on T cells after TCR stimulation. CD27 may be involved in cellular interaction of T and B lymphocytes.
Host Species: Mouse
Clone: M-T271
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
CD27 | |
Flow Cytometry | |
APC-H7 | |
RUO | |
Human | |
Human T-CLL cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
M-T271 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.