Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Hamster anti-Mouse, PerCP-Cy5.5, Clone: LG.3A10, BD

Catalog No. BDB563603 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563603 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563603 Supplier BD Biosciences Supplier No. 563603
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The LG.3A10 monoclonal antibody specifically binds to CD27, a lymphocyte-restricted member of the Tumor Necrosis Factor Receptor family which binds to CD70. The CD27 molecule is a 45-kDa transmembrane glycoprotein which is constitutively expressed by lymphocytes of the T lineage: virtually all thymocytes and over 90% of peripheral T cells bearing both αβ and γδ T-cell receptors. CD27 cooperates with the pre-TCR in mediating thymocyte differentiation and expansion. In addition, one to ten percent of mature peripheral B cells express CD27, and CD27’s role in the differentiation of human plasma cells has been studied. Mouse NK cells, freshly isolated and IL-2-activated, also express CD27. In the bone marrow, CD27 is found on a progenitor population which provides short-term hematopoietic reconstitution. Cells of the myeloid lineage do not express CD27. Cross-linked LG.3A10 mAb has been reported to amplify the proliferative response of purified T lymphocytes to suboptimal mitogenic stimulation1 and to enhance NK-cell proliferation and IFN-γ production. In contrast, non-cross-linked LG.3A10 mAb inhibits CD3-induced pre-T cell development by interfering with the receptor-ligand interaction. This hamster mAb to a mouse leukocyte antigen has been observed to cross-react with a similar population of rat leukocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD27
Applications Flow Cytometry
Classification Monoclonal
Clone LG.3A10
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description Tnfrsf7; Tumor necrosis factor receptor superfamily member 7; Tp55; S152
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Armenian hamster fibroblast line ARHO12 transfected with mouse Cd27 cDNA
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.