Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, PE, Clone: M-T271, BD

Catalog No. BDB557330 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557330 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557330 Supplier BD Biosciences Supplier No. 557330
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone M-T271 reacts with the human form of 110 kD disulfide-linked dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNFR/NGFR family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. This clone also cross-reacts with a major subset of peripheral blood lymphocytes and monocytes of baboon, and both rhesus and cynomolgus macaque monkeys. CD27+ cells were detected on all CD3+ lymphocytes and on a subset of CD20+ cells. The reactivity on lymphocytes and monocytes is similar to that observed with peripheral blood lymphocytes from normal human donors.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD27
Applications Flow Cytometry
Classification Monoclonal
Clone M-T271
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human T-CLL cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.