Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD270 (HVEM) Rat anti-Mouse, Alexa Fluor 647, Clone: C46, BD

Catalog No. BDB564470 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564470 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564470 Supplier BD Biosciences Supplier No. 564470

Rat Monoclonal Antibody

The C46 monoclonal antibody specifically binds to HVEM (Herpes virus entry mediator) which is also known as CD270. CD270 is a type I transmembrane protein and member of the TNF Receptor superfamily. It is encoded by Tnfrsf14 (tumor necrosis factor receptor superfamily member 14). CD270 is expressed by a variety of cell types including T cells, B cells, monocytes, dendritic cells, neutrophils, and mucosal epithelial cells. CD270 is a multifunctional factor that can both augment or inhibit immune responses. CD270 binds several ligands including CD258/LIGHT (is homologous to lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes), CD160, and CD272/BTLA. CD270-LIGHT interactions reportedly transduce costimulatory signals for T cells whereas CD270 interaction with BTLA can induce coinhibitory signals that regulate T cell-dependent immune responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD270 (HVEM)
Applications Flow Cytometry
Classification Monoclonal
Clone C46
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description HVEM; Herpes virus entry mediator; Tnfrsf14; HveA; Atar
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse HVEM Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.