Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD271 Mouse anti-Human, V450, Clone: C40-1457, BD

Catalog No. BDB562123 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562123 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562123 Supplier BD Biosciences Supplier No. 562123

Mouse Monoclonal Antibody

The C40-1457 monoclonal antibody specifically binds to CD271, the nerve growth factor receptor (NGFR). CD271 is a transmembrane 75 kDa protein that has been found localized to neuronal axons, Schwann cells, and perineural cells of peripheral nerves. It is also expressed by cells in some epithelial, mesenchymal and lymphoid tissues. NGFR is the receptor for nerve growth factor (NGF), apolypeptide that is essential for normal development of the nervous system. NGF promotes survival and differentiation of sympathetic and sensory neurons during embryological development of peripheral neurons. NGF binds to two distinctive surface receptors expressed by target cells, the p140-TrkA (NTRK1) and the p75 NGFR. High affinity binding of NGF requires that both receptor molecules be expressed. NGFR has been found on human and rat lymphocytes. A subset of lymphoid cells in the spleen, lymph nodes, and follicular dentritic cells in germinal centers of reactive lymph nodes were found to express CD271. It has been reported that NGFR interaction with its ligand, NGF, may play a role in immunoregulation. NGF may function as a B-cell growth factor.

The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD271
Applications Flow Cytometry
Classification Monoclonal
Clone C40-1457
Conjugate Violet Fluor 450 (V450)
Description NGFR; NTR; p75NTR; p75 ICD; TNFRSF16; TNR16; LNGFR
Formulation Aqueous buffered solution containing protein stabilizer, glycerol and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.