Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD272 Mouse anti-Human, PE, Clone: J168-540.90.22, BD

Catalog No. BDB558485 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558485 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558485 Supplier BD Biosciences Supplier No. 558485
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The J168-540 monoclonal antibody specifically binds to human CD272, also known as BTLA (B and T lymphocyte attenuator), an inhibitory receptor expressed in bone marrow and thymus on developing B and T cells. In the periphery, BTLA is expressed by B cells, T cells, bone marrow-derived dendritic cells, and macrophages. After T cell activation, BTLA appears to be expressed at higher levels on Th1 cell populations than in Th2 cell populations. Upon binding the herpesvirus entry mediator (HVEM/LIGHT-R/CD270), CD272 undergoes tyrosine phosphorylation and inhibits T cell proliferation in a BTLA-dependent manner. CD272 has structural similarities to two other lymphocyte inhibitory receptors, CTLA-4 and PD-1 and is a member of the CD28-like family of coreceptors. Based on these observations, CD272 is considered to be a negative regulator of lymphocyte activation and/or function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD272
Applications Flow Cytometry
Classification Monoclonal
Clone J168-540.90.22
Concentration 0.2mg/mL
Conjugate PE
Description BTLA1; B- and T-lymphocyte-associated protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BTLA Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.