Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD273 Rat anti-Mouse, APC, Clone: TY25, BD

Catalog No. BDB560086 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560086 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560086 Supplier BD Biosciences Supplier No. 560086
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TY25 monoclonal antibody specifically binds to CD273, also known as B7-DC or PD-L2. CD273 is a 42-kDa type I membrane glycoprotein encoded by the Pdcd1lg2 gene of the B7 family of the Ig superfamily. Pdcd1lg2 mRNA is expressed selectively in dendritic cells (DC), liver, and a few transformed cell lines (hepatic, neuroblastoma, and myeloid), but not in lymphoid tissues or macrophages. The protein has not been detected on resting peripheral T and B lymphocytes, macrophages, or DC, but it is upregulated upon activation of T cells, macrophages, and DC by a variety of stimulatory factors. B7-DC's receptor, PD-1, contains an ( I mmunoreceptor T yrosine-based I nhibitory M otif) on its intracytoplasmic region and is expressed on activated B and T lymphocytes, suggesting that B7-DC-PD-1 interaction may be involved in the negative regulation of immune responses. The second PD-1 ligand, B7-H1 (PD-L1), is also a member of the B7 family of the Ig superfamily. B7-DC may also participate in positive immunoregulation, or costimulation of T cells, through an additional receptor, which is not PD-1 and distinct from the alternate receptor for B7-H1. Moreover, B7-DC is the first B7 ligand that has been found to participate in bi-directional communication; ligation of B7-DC on DC stimulates those DC. The TY25 antibody blocks the binding of B7-DC to PD-1, but not to its alternate ligand.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD273
Applications Flow Cytometry
Classification Monoclonal
Clone TY25
Concentration 0.2mg/mL
Conjugate APC
Description Pdcd1lg2; B7dc; B7-DC; PD-1 ligand 2; PD-L2; Btdc
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse B7-DC
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.