Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD273 Mouse anti-Human, Brilliant Violet 650, Clone: MIH18, BD

Catalog No. BDB563844 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563844 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563844 Supplier BD Biosciences Supplier No. 563844
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH18 monoclonal antibody specifically binds to CD273 which is also known as the Programmed Death Ligand 2 (PD-L2) and B7-DC. CD273 is a type 1 transmembrane glycoprotein member of the B7 family and Ig superfamily. CD273 serves as a ligand for CD279, the Programmed Death 1 (PD-1) receptor. CD273 is expressed by dendritic cells, activated monocytes, medullary thymic epithelial cells, placental trophoblasts, and myocardial endothelium. CD273 can function as a coinhibitor of T cell functions by acting through CD279. Monoclonal antibodies that block the CD273 and CD279 interaction result in enhanced T cell proliferation and cytokine production. Thus, CD273 can play an important role in regulating T cell responses. Clone MIH18 is reportedly a blocking antibody.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon™ BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will bespillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD273
Applications Flow Cytometry
Classification Monoclonal
Clone MIH18
Conjugate Brilliant Violet 650
Description PD-1 ligand 2; PD-L2; B7-DC; PDCD1LG2; PDCD1 ligand 2; PDCD1L2; PDL2; Btdc
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD273 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.