Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD273 Mouse anti-Human, PerCP-Cy5.5, Clone: MIH18, BD

Catalog No. BDB564256 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564256 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564256 Supplier BD Biosciences Supplier No. 564256
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH18 monoclonal antibody specifically binds to CD273 which is also known as the Programmed Death Ligand 2 (PD-L2) and B7-DC. CD273 is a type 1 transmembrane glycoprotein member of the B7 family and Ig superfamily. CD273 serves as a ligand for CD279, the Programmed Death 1 (PD-1) receptor. CD273 is expressed by dendritic cells, activated monocytes, medullary thymic epithelial cells, placental trophoblasts, and myocardial endothelium. CD273 can function as a coinhibitor of T cell functions by acting through CD279. Monoclonal antibodies that block the CD273 and CD279 interaction result in enhanced T cell proliferation and cytokine production. Thus, CD273 can play an important role in regulating T cell responses. Clone MIH18 is reportedly a blocking antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD273
Applications Flow Cytometry
Classification Monoclonal
Clone MIH18
Conjugate PerCP-Cyanine5.5
Description PD-1 ligand 2; PD-L2; B7-DC; PDCD1LG2; PDCD1 ligand 2; PDCD1L2; PDL2; Btdc
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Human CD273 Transfected Cell Line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.