Learn More
BioLegend™ anti-CD274, Clone: 10F.9G2
Description
Host Species: Rat
Clone: 10F.9G2
Isotype: IgG2b, κ
Cross Reactivity: Mouse
Flow Cytometry
Specifications
Specifications
| Antigen | CD274 |
| Classification | Monoclonal |
| Clone | 10F.9G2 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Host Species | Rat |
| Purification Method | Unpurified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Target Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.