Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD275 Mouse anti-Human, Brilliant Violet 421, Clone: 2D3/B7-H2, BD

Catalog No. BDB564276 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564276 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564276 Supplier BD Biosciences Supplier No. 564276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2D3 monoclonal antibody specifically binds to CD275, which is also known as B7 homolog 2 (B7-H2/B7H2) and B7-related protein 1 (B7RP-1). CD275 is a type I transmembrane glycoprotein that belongs to the Ig superfamily. CD275 is likewise known as Inducible T-cell co-stimulator ligand (ICOS Ligand /ICOSLG/ICOS-L). It can bind to CD278 (ICOS) and costimulate the proliferation and cytokine production of antigen-stimulated human T cells. CD275 is variable expressed by subsets of B cells, monocytes, macrophages, dendritic cells and endothelial cells.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD275
Applications Flow Cytometry
Classification Monoclonal
Clone 2D3/B7-H2
Conjugate Brilliant Violet 421
Description ICOSLG; ICOS ligand; LICOS; B7-H2; B7 homolog 2; B7RP-1; GL50
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Host Species Mouse
Immunogen Human B7-H2
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.