Learn More
Description
Host Species: Rat
Clone MIH32
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Mouse B7-H3 Transfected Cell Line
Flow Cytometry
Specifications
Specifications
| Antigen | CD276 |
| Classification | Monoclonal |
| Clone | MIH32 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 488 |
| Host Species | Rat |
| Immunogen | Mouse B7-H3 Transfected Cell Line |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.