Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD278, BB515; Clone: 7E.17G9; BD

Catalog No. BDB564592 Shop All BD Biosciences Products
Change view
Click to view available options
Conjugate:
Brilliant Blue 515 (BB515)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
BDB564592 Brilliant Blue 515 (BB515)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564592 Supplier BD Biosciences Supplier No. 564592
Add to Cart
Add to Cart

Applications:

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD278
Applications Flow Cytometry
Classification Monoclonal
Clone 7E.17G9
Concentration 0.2mg/mL
Conjugate Brilliant Blue 515 (BB515)
Host Species Rat
Immunogen Mouse Icos cDNA and ICOS hexahistidine fusion protein
Purification Method Affinity Purified
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.