Learn More
CD278 Mouse anti-Human, Unlabeled, Clone: DX29, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 557801
Description
The DX29 monoclonal antibody specifically binds to human CD278 which is also known as inducible costimulator (ICOS) and inducible T-cell co-stimulator. ICOS is a homodimeric type I transmembrane glycoprotein with an approximate molecular weight of 50-60 kDa. It is a member of the CD28 family and is highly expressed on activated T cells. CD278 is the receptor for ICOS-ligand (also known as CD275, B7-H2, B7RP-1, and LICOS). Like CD28, ICOS can provide a costimulatory signal for T cell activation, proliferation and cytokine production. It is not expressed on resting or activated B cells, monocytes, NK cells, granulocytes, dendritic cells or platelets. Unlike the constitutively expressed CD28, ICOS is de novo expressed upon cellular activation. Reports describe similarities between CD28 and ICOS in T cell activation, such as the costimulation of cytokine production. However, it has been suggested that ICOS may play a greater role in IL-10 production. In the presence of IL-10, purified recombinant human ICOS protein significantly increased in vitro B cell growth stimulated by pokeweed mitogen (PWM) and enhanced production of IgG.

Specifications
CD278 | |
Monoclonal | |
0.5mg/mL | |
ICOS; DX-29; H4; Inducible T-cell costimulator; AILIM; CVID1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
DX29 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Activated human T cells | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.