Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD279 Mouse anti-Human, Brilliant Violet 650, Clone: EH12.1, BD

Catalog No. BDB564104 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564104 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564104 Supplier BD Biosciences Supplier No. 564104
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The EH12.1 monoclonal antibody specifically binds to CD279. CD279 is an immunoregulatory receptor that is expressed on activated T cells, B cells and myeloid cells and contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279
Applications Flow Cytometry
Classification Monoclonal
Clone EH12.1
Conjugate Brilliant Violet 650
Description hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PD-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.