Learn More
CD279 (PD-1) Mouse anti-Human, NA/LE, Unlabeled, Clone: EH12.1 (also known as EH12), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562138
Description
The EH12.1 monoclonal antibody specifically binds to CD279. CD279 is an immunoregulatory receptor that is expressed on activated T cells, B cells and myeloid cells and contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) in the cytoplasmic region. Mice deficient in CD279 show a breakdown of peripheral tolerance and manifest multiple autoimmune symptoms. PD-L1 and PD-L2 are ligands of CD279 and are members of the B7 gene family. Interaction of CD279:PD-Ligands results in inhibition of T cell proliferation and cytokine secretion. Reports suggest that the B7/CTLA-4 pathway functions primarily to attenuate, limit, and/or terminate naοve T-cell activation in secondary lymphoid organs. The PD-ligand:CD279 pathway, on the other hand, may primarily attenuate, limit, and/or terminate T-, B-, and myeloid cell activation/effector function at sites of inflammation in the periphery.

Specifications
CD279 (PD-1) | |
Monoclonal | |
1.0mg/mL | |
hPD-1; PD1; PDCD1; Programmed cell death 1; SLEB2 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
EH12.1 (also known as EH12) | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Human PD-1 Recombinant Protein | |
0.5 mg | |
Primary | |
Store undiluted at 4°C |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.