Learn More
anti-CD28, Alexa Fluor 488 , Clone: 794716, R&D Systems™
CD28
Supplier: R&D Systems FAB4832G
Description
CD28 and CTLA-4, together with their ligands, B7-1 and B7-2, constitute one of the dominant costimulatory pathways that regulate T and B cell responses. CD28 and CTLA-4 are structurally homologous molecules that are members of the immunoglobulin (Ig) gene superfamily. Mouse CD28 is expressed constitutively on virtually 100% of mouse Tcells and on developing thymocytes.
Host Species: Rat
Clone: 794716
Isotype: IgG1
Species Reactivity: Ms
Immunogen: Mouse myeloma cell line NS0-derived recombinant mouse CD28; Accession Number P31041
Applications:
Flow Cytometry
Order Info
The product is shipped with polar packs.
Specifications
CD28 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD28, CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
940 | |
Mouse | |
Purified |
Flow Cytometry | |
794716 | |
Flow Cytometry 5 uL/10^6 cells | |
P31041 | |
CD28 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD28, Accession # P31041 | |
100 Tests | |
Primary | |
Detects mouse CD28 in ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.