Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, Clone: 15E8, MilliporeSigma™

Catalog No. CBL517MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL517MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL517MI Supplier MilliporeSigma Supplier No. CBL517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-CD28 Antibody, clone 15E8 is validated for use in flow cytometry, FUNC for the detection of CD28.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 15E8
Conjugate Unconjugated
Formulation Please see lot specific datasheet for product formulation. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Gene Accession No. NM_006139.1
Gene Symbols CD28, Tp44, TP44, MGC138290
Host Species Mouse
Immunogen Human T lymphocytes
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for up to one year.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.