Learn More
CD28 Mouse anti-Human, Brilliant Violet 421, Clone: CD28.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 562613
Description
The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications
CD28 | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD28 Transfected Cell Line | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
CD28.2 | |
CD28 antigen; T44; Tp44; TP44 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.