Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Rat, NA/LE, Unlabeled, Clone: JJ316, BD

Catalog No. BDB554992 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554992 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554992 Supplier BD Biosciences Supplier No. 554992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JJ316 antibody reacts with CD28 on virtually all T lymphocytes bearing αβT-cell receptors (TCR), on most γδTCR-bearing T cells, and on a subset of NK cells. In the thymus, CD28 expression is developmentally regulated during the maturation of αβTCR-bearing T cells. CD28 is a costimulatory receptor required for activation of T cells; its ligands include CD80 (B7-1) and CD86 (B7-2). Soluble JJ316 enhances the proliferative responses and IL-2 production by T cells activated by anti-aß-TCR mAb R73 (Cat. no. 554910). Furthermore, at high concentrations, cross-linked JJ316 mAb stimulates resting CD4+ and CD8+T cells to proliferate, secrete IL-2, express activation markers, and gain CTL activity in the absence of signaling through the TCR. In vivo administration of mAb JJ316 results in blastogenic transformation of peripheral CD4+ cells and dramatic lymphocytosis. The alternate anti-rat CD28 mAb JJ319 (Cat. no. 554993) cannot directly activate T cells, although it has in vitro costimulatory activity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone JJ316
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Rat Cd28-transfected BALB/c mouse A20J B lymphoma
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.