Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Rat, Unlabeled, Clone: JJ319, BD

Catalog No. BDB559982 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559982 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559982 Supplier BD Biosciences Supplier No. 559982
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The JJ319 antibody reacts with CD28, which is expressed on virtually all T lymphocytes bearing αβ T-cell receptors (TCR), on most γδ TCR-bearing T cells, and on a subset of NK cells. In the thymus, CD28 expression is developmentally regulated during the maturation of αβ TCR-bearing T cells. CD28 is a costimulatory receptor required for activation of T cells; its ligands include CD80 (B7-1) and CD86 (B7-2). Soluble JJ319 mAb costimulates the proliferative responses and IL-2 production of CD4+ and CD8+ T cells activated by anti-αβ-TCR mAb R73 (Cat. no. 554910). The alternate anti-rat CD28 mAb JJ316 (Cat. No. 554992) is capable of directly stimulating T cells in vitro and in vivo.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone JJ319
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat CD28-transfected cell line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.