Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, PE, Clone: L293, BD

Catalog No. BDB348047 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB348047 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB348047 Supplier BD Biosciences Supplier No. 348047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD28 antibody, clone L293, is derived from hybridization of Sp2/0-Ag14 mouse myeloma cells with spleen cells from BALB/c mice immunized with the HPB-ALL T-cell line.

The CD28 antigen, a disulfide-linked homodimeric glycoprotein, Mr 44 kilodaltons (kd), is a cell-adhesion molecule (CAM) and functions as the ligand for CD80 (B7-1) and CD86 (B7-2) antigens, which are present on activated B lymphocytes, monocytes, and dendritic cells. Interaction of the CD28 antigen with CD80 or CD86 antigens, or both, co-stimulates CD2 and CD3 antigen/T-cell antigen receptor (TCR)–dependent T-cell–mediated proliferation and cytotoxicity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD28
Applications Flow Cytometry
Classification Monoclonal
Clone L293
Concentration 25μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.