Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD284/MD-2 Complex Rat anti-Mouse, PE, Clone: MTS510, BD

Catalog No. BDB558294 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558294 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558294 Supplier BD Biosciences Supplier No. 558294
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MTS510 antibody reacts with the molecular complex of Toll-Like Receptor 4 and MD-2 (TLR4-MD-2) which is expressed on LPS responsive macrophages. TLR4, a member of the Toll-Like Receptor Family, has been renamed as CD284 and identified to be the transmembrane signal-transducing portion of the receptor for LPS. TLR4 associates on the cell surface with CD14 and MD-2, a 0.7 kDa molecule which is anchored to the membrane via its physical association with TLR4. The association of MD-2 with TLR4 is required for recognition of LPS and the anti-mitotic compound Taxol, which mimics the action of LPS on mouse cells. MTS510 mAb detects TLR4-MD-2 on the surface of thioglycollate-elicited macrophages from all mouse strains tested (ie, BALB/c, C57BL/6, C3H/HeJ, C3H/HeN, and DBA/1), including the C3H/HeJ strain which expresses an LPS-resistant mutant TLR4. Expression of TLR4-MD-2 is down-regulated on peritoneal macrophages after exposure to LPS, correlating with the occurrence of LPS tolerance. TLR4-MD-2 is not detected on splenocytes or thymocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD284/MD-2 Complex
Applications Flow Cytometry
Classification Monoclonal
Clone MTS510
Concentration 0.2mg/mL
Conjugate PE
Description TLR4/MD-2 Complex
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Pro-B cell line Ba/F3 e pressing transfected BALB/c mouse TLR4 and MD-2
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.