Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 Mouse anti-Human, APC, Clone: MAR4, BD

Catalog No. BDB559883 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559883 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559883 Supplier BD Biosciences Supplier No. 559883
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAR4 monoclonal antibody specifically binds to CD29. CD29 is a 130 kDa integrin β1 subunit that is expressed as a heterodimeric complex with one of six distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The β1 subunit has a broad tissue distribution; it is expressed on lymphocytes, monocytes and weakly on granulocytes, but not on erythrocytes. These receptors are involved in a variety of cell-cell and cell-matrix interactions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29
Applications Flow Cytometry
Classification Monoclonal
Clone Mar4
Conjugate APC
Description ITGB1; Integrin beta-1; Integrin β1; FNRB; GPIIA; ITB1; MDF2; MSK12; VLA-4β
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Human α5β1 Fibronectin Receptor
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.