Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 Mouse, Unlabeled, Clone: 18, BD

Catalog No. BDB610468 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610468 150 μg
BDB610467 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610468 Supplier BD Biosciences Supplier No. 610468
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrins are membrane receptors that mediate cell-cell or cell-matrix adhesion. All integrins are transmembrane heterodimers composed of α and β subunits and are connected to the cytoskeleton. At least 20 integrins, formed from combinations of 12 α and 9 β subunits, have been reported. Many of these have been implicated as transducers of molecular signals. The β1 subgroup of this receptor family comprises at least six different dimer combinations. Among these combinations, α2β1 is associated with type I collagen and laminin binding and regulation, α3β1 is a receptor for laminin and epiligrin, and α5β1 is a fibronectin receptor. β1 integrins play an important role in several aspects of epidermal differentiation and morphogenesis. Expression of the β1 subunit is regulated by growth factors such as TGF-β1. Integrin activation, which enhances binding of T cells to endothelium, is regulated in part by phosphatidylinositol 3-kinase. Focal adhesion kinase (FAK) and paxillin have been reported to independently bind the C-terminal, cytoplasmic domain of the β1 subunit. FAK is reported to be enzymatically activated upon engagement of integrins with their ligands and paxillin is phosphorylated on tyrosine upon activation of FAK.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Description Integrin β1; platelet GPIIa; VLA β-chain
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.