Learn More
CD29 Hamster anti-Rat, NA/LE, Unlabeled, Clone: HA2/5, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 555002
Description
The Ha2/5 antibody reacts with the 130kDa integrin β1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin α chains. With α1 through α6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with αV (CD51), it forms αVβ1 integrin. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, and epithelia. The Ha2/5 hamster anti-rat CD29 monoclonal antibody cross-reacts with mouse thymocytes, splenocytes, and peripheral lymph node leukocytes. The Ha2/5 antibody blocks in vitro adhesion of CD29-expressing cells to collagen.

Specifications
CD29 | |
Monoclonal | |
1.0mg/mL | |
Itgb1; Integrin β1 chain; Integrin beta-1; VLA-4 subunit beta | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
HA2/5 | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Rat glomerular epithelial cells | |
0.5 mg | |
Immunology | |
Mouse, Rat | |
IgM κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.