Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 Rat anti-Mouse, Unlabeled, Clone: KMI6, BD

Catalog No. BDB558741 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558741 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558741 Supplier BD Biosciences Supplier No. 558741
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The KMI6 antibody reacts with the 130-kDa integrin ß1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin achains. With a1 through a6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with av (CD51), it forms avß1 integrin. It also associates with the integrin a7, a8, and a9 chains in non-lymphoid tissues. As a result, CD29 has a broad tissue distribution, including leukocytes, endothelia, smooth muscle, epithelia, and oocytes. The epitope recognized by the KMI6 mAb can be detected by immunoblot in unreduced extracts from a variety of cells and tissues having a number of different ß1-integrin heterodimer combinations. However, the epitope is not well expressed on the surface of the majority of hematopoietic cells. It is detected on the surface of fetal liver cells, some bone marrow stromal cells, and some cultured hematopoietic cell lines, including P815 and PA6. Surface expression of the CD29 epitope recognized by mAb KMI6 on thymocytes and T lymphocytes appears to be developmentally regulated, and KMI6 antibody enhances the adhesion of KMI6 thymocytes to fibronectin and laminin.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29
Applications Flow Cytometry
Classification Monoclonal
Clone KMI6
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Integrin β1 chain
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen (C57BL/6 x DBA/2)F1 mouse bone-marrow stromal cell clone BMS2
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.