Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD29 Hamster anti-Rat, PE, Clone: Ha2/5, BD

Catalog No. BDB562154 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562154 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562154 Supplier BD Biosciences Supplier No. 562154

Hamster Monoclonal Antibody

The Ha2/5 monoclonal antibody specifically binds to the 130 kDa integrin β1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin α chains. With α1 through α6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with αV (CD51), it forms αVβ1 integrin. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, and epithelia. The Ha2/5 hamster anti-rat CD29 monoclonal antibody cross-reacts with mouse thymocytes, splenocytes, and peripheral lymph node leukocytes. The Ha2/5 antibody blocks in vitro adhesion of CD29-expressing cells to collagen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29
Applications Flow Cytometry
Classification Monoclonal
Clone Ha2/5
Concentration 0.2mg/mL
Conjugate PE
Description Itgb1; Integrin β1 chain; Integrin beta-1; VLA-4 subunit beta
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Rat glomerular epithelial cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.