Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD294 Rat anti-Human, Unlabeled, Clone: BM16, BD

Catalog No. BDB558412 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558412 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558412 Supplier BD Biosciences Supplier No. 558412
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BM16 monoclonal antibody specifically binds to CD294. CD294 is encoded by PTGDR2 (Prostaglandin D2 receptor 2) and is also known as CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells), GPR44 (G protein-coupled receptor 44), DL1R, and DP2. CD294 is a member of the G protein-coupled leukocyte chemoattractant receptor family and it is expressed on Th2, but not Th1 type cells. CD294 is detectable on CD4+ T cells in fresh PBMC but not on B cells and NK cells. CD294 is also expressed on peripheral blood basophils and eosinophils, suggesting its involvement allergic reactions. Phenotypic analysis of CD4+ T cells expressing CRTH2 demonstrated that they were also CD45RA-negative and CD45RO+ and CD25+. These cells produce Th2- but little or no Th1-type cytokines upon stimulation with PMA and Ionomycin.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD294
Applications Flow Cytometry
Classification Monoclonal
Clone BM16
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CRTH2; PTGDR2 ; Prostaglandin D2 receptor 2; DL1R; DP2; GPR44
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Transfectant cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.