Learn More
BioLegend™ anti-CD3ε, Clone: 145-2C11
Supplier: BioLegend™ 100306
Description
Host Species: Hamster
Clone: 145-2C11
Isotype: IgG
Cross Reactivity: Mouse
Immunogen: H-2Kb-specific mouse cytotoic T lymphocyte clone BM10-37
Applications:
Flow Cytometry
Specifications
| CD3 ε | |
| 145-2C11 | |
| FITC | |
| H-2Kb-specific mouse cytotoic T lymphocyte clone BM10-37 | |
| 500 μg | |
| Mouse |
| Monoclonal | |
| 0.5mg/mL | |
| Hamster | |
| Unpurified | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.