Learn More
CD3ε Hamster anti-Mouse, LE/AF (Low Endotoxin/Azide Free), Clone: 145-2C11, Southern Biotech™
Hamster Monoclonal Antibody
Supplier: Southern Biotechnology Associates 153014
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD3ε | |
Low Endotoxin/Azide-free (LE/AF) | |
0.5mg/mL | |
Hamster | |
0.5 mg | |
Primary | |
Store at -20°C |
Blocking Assay, Depletion Assay, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
145-2C11 | |
Supplied in PBS | |
H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone BM10-37 | |
RUO | |
Mouse | |