Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3ε Hamster anti-Mouse, FITC, Clone: 500-A2, Southern Biotech™

Hamster Monoclonal Antibody
Supplier: Southern Biotechnology Associates 153102
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| CD3ε | |
| Monoclonal | |
| 0.5mg/mL | |
| Supplied in PBS/NaN3 | |
| Immune complexes isolated from C6VL-BS thymoma cells | |
| RUO | |
| Mouse | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| 500-A2 | |
| FITC | |
| Hamster | |
| 0.5 mg | |
| Primary | |
| Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction