Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Human, Unlabeled, Clone: SP34, BD

Catalog No. BDB556610 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556610 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556610 Supplier BD Biosciences Supplier No. 556610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone SP34 reacts with human form of the 20 kDa epsilon subunit of the CD3 antigen/T-cell receptor (TCR) complex. This clone also cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of rhesus or cynomolgus and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes, with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cell markers.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone SP34
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C
Isotype IgG3 λ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.