Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Human, Unlabeled, Clone: UCHT1, BD

Catalog No. BDB555330 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555330 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555330 Supplier BD Biosciences Supplier No. 555330
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Studies from the HLDA Workshop show this antibody to be mitogenic when used in conjunction with pokeweed mitogen. CD3 plays a role in signal transduction during antigen recognition. UCHT1 antibody stains intracellular CD3 unlike the other CD3 clone, HIT3a, which stains only the extracellular CD3. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone UCHT1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.