Learn More
BioLegend™ CD3, Mouse anti-Human, Cy5.5. PerCP, Clone: UCHT1
Specifications
Specifications
| Antigen | CD3 |
| Classification | Monoclonal |
| Clone | UCHT1 |
| Conjugate | PerCP-Cyanine5.5 |
| Host Species | Mouse |
| Purification Method | Unpurified |
| Quantity | 100 Tests |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.