Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3ε Hamster anti-Mouse, FITC, Clone: 145-2C11, BD

Catalog No. BDB553062 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553062 0.5 mg
BDB553061 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB553062 Supplier BD Biosciences Supplier No. 553062
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The 145-2C11 antibody reacts with the 25-kDa ε chain of the T-cell receptor-associated CD3 complex, which is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events which activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes and it has been reported that pre-incubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Hamster
Clone: 145-2C11
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: H-2Kb specific cytotoxic T lymphocyte clone BM10-37

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 ε
Applications Flow Cytometry
Classification Monoclonal
Clone 145-2C11
Concentration 0.5mg/mL
Conjugate FITC
Description CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.