Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3ε Mouse anti-Human, FITC, Clone: APA1/1, BD

Catalog No. BDB558261 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558261 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558261 Supplier BD Biosciences Supplier No. 558261

Mouse Monoclonal Antibody

Monoclonal antibody APA1/1 reacts with the intracellular domain of the epsilon chain of the CD3 molecule (CD3- ε ). This antibody does not react with the extracellular region of CD3- ε . The assembly of the T cell antigen receptor (TCR)-CD3 complex has been suggested to take place by pairwise interactions of the CD3- ε subunit with either CD3- γor CD3- δ . These dimers then associate with the TCR heterodimer a/ β or γε . Other anti-CD3 antibodies (OKT3, WT31, UCHT1, Leu4) did not react with COS cells singly transfected with CD3- ε . This reagent could be useful for the study of T cell development or the study of conformational changes of CD3- ε upon ligand binding to TCR-CD3 complex.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 ε
Applications Flow Cytometry
Classification Monoclonal
Clone APA1/1
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.