Learn More
CD3 Mouse anti-Human, FITC, Clone: UCHT1, BD
Mouse Monoclonal Antibody
$62.00 - $552.50
Specifications
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561806
|
BD Biosciences
561806 |
25 Tests |
Each for $62.00
|
|
|||||
BDB561807
|
BD Biosciences
561807 |
500 Tests |
Each for $552.50
|
|
|||||
Description
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
Specifications
CD3 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon |
UCHT1 | |
Monoclonal | |
Mouse | |
Human | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.