Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD3 Hamster anti-Mouse, FITC, Clone: 145-2C11, MilliporeSigma™
Hamster Monoclonal Antibody
Supplier: MilliporeSigma FCMAB227F
Description
This Milli-Mark™ Anti-mouse CD3-FITC Antibody, clone 145-2C11 is validated for use in FC for the detection of mouse CD3.
Specifications
| CD3 | |
| Monoclonal | |
| FITC | |
| NP_031674.1. | |
| Hamster | |
| Protein G Purified | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 6 months. | |
| IgG |
| Flow Cytometry | |
| 145-2C11 | |
| Purified hamster monoclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA. | |
| Cd3e | |
| Animals were immunized with the BM10-37 mouse cytotoxic T lymphocyte (CTL) cell clone (anti H-2 Kb). Spleen cells were fused with Sp2/0-Ag14 myeloma cells. | |
| 100 Tests | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction