Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD3, Polyclonal

Catalog No. PC630100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC630100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC630100UL Supplier MilliporeSigma™ Supplier No. PC630100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects CD3 Clone: in Human, Mouse samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

The CD3 antigen is present in early thymocytes, and has been considered the best all-around T cell marker. The positive staining of this marker may represent a sign of early commitment to the T cell lineage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In proprietary buffer.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids from human CD3
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes CD3 present in early thymocytes.
Target Species Human, Mouse
Content And Storage 2°C to 8°C
Form Diluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.