Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Human, V500, Clone: UCHT1, BD

Catalog No. BDB561417 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561417 25 Tests
BDB561416 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561417 Supplier BD Biosciences Supplier No. 561417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.

The antibody is conjugated to BD Horizon™ V500, which has been developed for use in multicolor flow cytometry experiments and is

available exclusively from BD Biosciences. It is excited by the Violet laser with an Ex max of 415nm and Em Max at 500 nm. BD Horizon V500 conjugates emit at a similar wavelength to AmCyan yet exhibit reduced spillover into the FITC channel. For more information on BD Horizon V500, visit bdbiosciences.com/colors.

When compensating dyes in this spectral range (such as Horizon™ V500 and AmCyan), the most accurate compensation can be obtained using single stained cellular controls. Due to spectral differences between cells and beadsin this channel, using BD CompBeads can result in spillover errors for V500 and AmCyan reagents.Therefore, the use of BD CompBeads or BD CompBeads Plus to determine spillover values for these reagents is not recommended.Different V500 reagents (e.g., CD4 vs. CD45) can have slightly different fluorescence spillover levels. Therefore, it may also be necessary to use clone specific compensation controls when using these reagents.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone UCHT1
Conjugate Violet Fluor 500 (V500)
Description CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon
Formulation Aqueous buffered solution containing protein stabilizer, glycerol and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human infant thymocytes and peripheral blood lymphocytes from a Sezary Syndrome donor
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.