Learn More
CD3z Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137284
Description
This antibody is predicted to react with bovine and porcine based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is tonsil tissue.
CD3 (Cluster of Differentiation 3) consists of five invariant polypeptide chains that associate to form three dimmers: a heterodimer of gamma and epsilon chains, a heterodimer of delta and epsilon chains and a homodimer of two zeta chains or a heterodimer of zeta and eta chains. The zeta chain contains three consecutive copies of the same motif.
Specifications
CD3z | |
Polyclonal | |
CD247 | |
T3Z, CD3H, CD3Q, TCRZ, CD3-ZETA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
919 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P20963 | |
CD247 | |
Synthetic peptide derived from near N-terminus of the zeta chain of human CD3 protein. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.